Learn More
ACROBiosystems Human CD45 / PTPRC protein, His Tag
Supplier: ACROBiosystems CD5H52H8100UG
Recombinant Protein;100UG;Human CD45, His Tag (CD5-H52H8) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Lys 577 (Accession # P08575-3).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 62.6 kDa.