Learn More
ACROBiosystems Biotinylated Human IL-13 R alpha 2 Protein, His,Avitag™
Supplier: ACROBiosystems IL2H82E625UG
Recombinant Protein;25UG;Biotinylated Human IL-13 R alpha 2, His,Avitag (IL2-H82E6) is expressed from human 293 cells (HEK293). It contains AA Asp 27 - Arg 343 (Accession # Q14627-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 41.9 kDa.