Learn More
ACROBiosystems SARS-CoV-2 (COVID-19) S protein RBD (N439K), His Tag (MALS verified)
Manufacturer:  ACROBiosystems SPDC52HG100UG
Encompass_Preferred
Recombinant Protein;100UG;SARS-CoV-2 S protein RBD (N439K), His Tag (SPD-C52Hg) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (N439K)).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.5 kDa.
Catalog No. 50-205-4915