Learn More
ACROBiosystems Biotinylated Human CD133 Protein, His Tag (Nanodisc)
Manufacturer:  ACROBiosystems CD3H82E620UG
Encompass_Preferred
Recombinant Protein;20UG;Biotinylated Human CD133, His Tag (Nanodisc) (CD3-H82E6) is expressed from human 293 cells (HEK293). It contains AA Gly 20 - His 865 (Accession # O43490-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 98.9 kDa.
Catalog No. 50-205-4924