Learn More
ACROBiosystems Human HGF Protein, Fc Tag
Supplier:  ACROBiosystems HGFH5253100UG
Encompass_Preferred
Recombinant Protein;100UG;Human HGF, Fc Tag (HGF-H5253) is expressed from human 293 cells (HEK293). It contains AA Gln 32 - Ser 728 (Accession # P14210-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 106.1 kDa | 53.7 kDa | 52.4 kDa.
Catalog No. 50-205-4926