Learn More
ACROBiosystems Human GDF-2 Protein, Tag Free
Supplier:  ACROBiosystems GD2H521150UG
Encompass_Preferred
Recombinant Protein;50UG;Human GDF-2, Tag Free (GD2-H5211) is expressed from human 293 cells (HEK293). It contains AA Ser 320 - Arg 429 (Accession # Q9UK05-1).;This protein carries no "tag". The protein has a calculated MW of 12.1 kDa. The protein migrates as 14 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human GDF-2, Tag Free (Cat. No. GD2-H5211) at 1 μg/mL (100 μL/well) can bind Human ALK-1, His Tag (Cat. No. AL1-H5227) with a linear range of 2-16 ng/mL (QC tested).
Catalog No. 50-205-4934