Learn More
ACROBiosystems Anti-SARS-CoV-2 Spike RBD Neutralizing Antibody, Chimeric mAb, Human IgM (AM122)
Manufacturer: ACROBiosystems SPDM162100UG
Antibody;100UG;IgM/kappa;This product is a specific antibody against SARS-CoV-2 Spike protein RBD domain. No cross-reactivity is deteced with Spike protein RBD domain of other coronaviruses, including SARS-CoV, MERS-CoV, HCoV-229E, HCoV-NL63, HCoV-OC43 and HCoV-HKU1.;Anti-SARS-CoV-2 Spike RBD Neutralizing antibody, Human IgM (SPD-M162) is isolated from a SARS-CoV-2 infected patient and is recombinantly produced from human 293 cells (HEK293).; The protein migrates as 62.6 kDa and 23.5 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Anti-SARS-CoV-2 Spike RBD Neutralizing Antibody, Chimeric mAb, Human IgM (Cat. No. SPD-M162) at 2 μg/mL (100 μL/well) can bind SARS-CoV-2 S protein RBD, His Tag (Cat. No. SPD-C52H1) with a linear range of 0.6-2.4 ng/mL (QC tested).;Serial dilutions of Anti-SARS-CoV-2 Spike RBD Neutralizing Antibody, Chimeric mAb, Human IgM (Cat. No. SPD-M162) was detected by Biotinylated Human ACE2, His,Avitag (Cat. No. AC2-H82E6) with a half maximal inhibitory c
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.