Learn More
ACROBiosystems Cynomolgus Mucin-1 / MUC-1 Protein, His Tag
Supplier:  ACROBiosystems MU1C52H5100UG
Encompass_Preferred
Recombinant Protein;100UG;Cynomolgus Mucin-1, His Tag (MU1-C52H5) is expressed from human 293 cells (HEK293). It contains AA Leu 254 - Gly 373 (Accession # XP_005541632.1).;This protein carries a polyhistidine tag at the C-terminus.The mature form of Mucin-1 is a non-covalent heterodimeric complex with the proteolytically cleaved partial α and partial β chain. Each partial α and partial β chain has a calculated MW of 7.2 kDa (partial α chain) and 8.3 kDa (partial β chain). The protein migrates as 14 kDa (uncleaved), 12 kDa (partial α chain) and 11 kDa (partial β chain) under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-205-4941