Learn More
ACROBiosystems Human ENPP3 / CD203c Protein, His Tag
Supplier:  ACROBiosystems EN3H52H4100UG
Encompass_Preferred
Recombinant Protein;100UG;Human ENPP3, His Tag (EN3-H52H4) is expressed from human 293 cells (HEK293). It contains AA Leu 48 - Ile 875 (Accession # O14638-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 96.9 kDa.
Catalog No. 50-205-4951