Learn More
ACROBiosystems MERS S1 protein, His Tag
Supplier:  ACROBiosystems S1NM52H5100UG
Encompass_Preferred
Recombinant Protein;100UG;MERS S1 protein, His Tag (S1N-M52H5) is expressed from human 293 cells (HEK293). It contains AA Tyr 18 - Arg 751 (Accession # K0BRG7-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 83.1 kDa. The protein migrates as 90-116 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human DPPIV, Fc Tag (Cat. No. DP4-H5266) at 2 μg/mL (100 μL/well) can bind MERS S1 protein, His Tag (Cat. No. S1N-M52H5) with a linear range of 0.039-0.156 μg/mL (QC tested).
Catalog No. 50-205-4968