Learn More
ACROBiosystemsSupplier Diversity Partner Human FGF R2 (IIIc) protein, His Tag (MALS verified)  DFS Item
Manufacturer:  ACROBiosystemsSupplier Diversity Partner FGCH5225100UG
Recombinant Protein;100UG;Human FGFR2 (IIIc), His Tag (FGC-H5225) is expressed from human 293 cells (HEK293). It contains AA Pro 154 - Thr 358 (Accession # P21802-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.9 kDa. The protein migrates as 38-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human FGFR2 (IIIc), His Tag (Cat. No. FGC-H5225) at 5 μg/mL (100 μL/well) can bind Biotinylated Human FGF basic, Tag Free with a linear range of 0.04-1.25 μg/mL (Routinely tested).
Catalog No. 50-205-4979