Learn More
ACROBiosystemsSupplier Diversity Partner Cynomolgus FGF R2 (IIIb) protein, His Tag (MALS verified)
Manufacturer:  ACROBiosystemsSupplier Diversity Partner FGBC5225100UG
Encompass_Preferred
Recombinant Protein;100UG;Cynomolgus FGFR2 (IIIb), His Tag (FGB-C5225) is expressed from human 293 cells (HEK293). It contains AA Pro 144 - Thr 346 (Accession # A0A2K5TL84-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.6 kDa. The protein migrates as 35-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-205-4981