Learn More
ACROBiosystemsSupplier Diversity Partner Biotinylated SARS-CoV-2 (COVID-19) S protein RBD, Mouse IgG2a Fc,Avitag™(MALS verified)
Manufacturer:  ACROBiosystemsSupplier Diversity Partner SPDC82A925UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated SARS-CoV-2 S protein RBD, Mouse IgG2a Fc,Avitag (SPD-C82A9) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1).;This protein carries a mouse IgG2a Fc tag at the C-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 53.2 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human ACE2, Fc Tag (Cat. No. AC2-H5257) at 1 μg/mL (100 μL/well) can bind Biotinylated SARS-CoV-2 S protein RBD, Mouse IgG2a Fc,Avitag (Cat. No. SPD-C82A9) with a linear range of 0.2-6 ng/mL (QC tested).;Immobilized Biotinylated SARS-CoV-2 S protein RBD, Mouse IgG2a Fc,Avitag (Cat. No. SPD-C82A9) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) (0.5 μg/well) plate. can bind Anti-SARS-CoV-2 RBD Neutralizing Antibody, Human IgG1 (Cat. No. SAD-S35) with a linear range of 0.1-2 ng/mL (Routinely tested).
Catalog No. 50-205-5004