Learn More
ACROBiosystemsSupplier Diversity Partner Human Kremen-1 protein, Fc Tag  DFS Item
Manufacturer:  ACROBiosystemsSupplier Diversity Partner KR1H5253100UG
Recombinant Protein;100UG;Human Kremen-1, Fc Tag (KR1-H5253) is expressed from human 293 cells (HEK293). It contains AA Arg 21 - Thr 392 (Accession # Q96MU8-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 67.4 kDa.
Catalog No. 50-205-5014