Learn More
ACROBiosystems Human IL-2 R beta&IL-2 R alpha&IL-2 R gamma Protein, Fc Tag&Fc Tag (MALS verified)
Supplier:  ACROBiosystems ILGH5257100UG
Encompass_Preferred
Recombinant Protein;100UG;Human IL-2RB&IL-2RA&IL-2RG, Fc Tag&Fc Tag (ILG-H5257) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Asp 239 & Glu 22 - Ser 212 & Leu 23 - Asn 254 (Accession # P14784-1 (IL-2RB) & P01589-1 (IL-2RA) & P31785-1 (IL-2RG)).;Human IL-2RB&IL-2RA&IL-2RG, Fc Tag&Fc Tag is produced by co-expression of IL-2RB and IL-2RA and IL-2RG, has a calculated MW of 51.1 kDa (IL-2RB) and 75.4 kDa (IL-2RA&IL-2RG). Subunit IL-2RB is fused with a human IgG1 Fc tag at the C-terminus and subunit IL-2RA&IL-2RG is fused with a human IgG1 Fc tag at the C-terminus. The reducing (R) protein migrates as 60-66 kDa and 100-116 kDa respectively due to glycosylation.;Immobilized Biotinylated Human IL-2, His,Avitag (Cat. No. IL2-H82E4) at 1 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate can bind Human IL-2RB&IL-2RA&IL-2RG, Fc Tag&Fc Tag (Cat. No. ILG-H5257) with a linear range of 0.4-6 ng/mL (QC tested).;Human IL-2RB&IL-2RA&IL-2RG, Fc Tag&Fc Tag (Cat. No. IL
Catalog No. 50-205-5028