Learn More
ACROBiosystemsSupplier Diversity Partner Biotinylated Human GITR / TNFRSF18 Protein, His,Avitag™ (MALS verified)
Manufacturer:  ACROBiosystemsSupplier Diversity Partner GIRH82E125UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human GITR, His,Avitag™ (GIR-H82E1) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Glu 161 (Accession # Q9Y5U5-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 18.1 kDa. The protein migrates as 22-26 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human GITR Ligand, Fc Tag (Cat. No. GIL-H5261) at 10 μg/mL (100 μL/well) can bind Biotinylated Human GITR, His,Avitag (Cat. No. GIR-H82E1) with a linear range of 0.2-8 ng/mL (QC tested).
Catalog No. 50-205-5030