Learn More
ACROBiosystemsSupplier Diversity Partner Anti-SARS-CoV-2 RBD Potent Neutralizing Antibody, Chimeric mAb, Human IgG1 (AM128)  DFS Item
Manufacturer:  ACROBiosystemsSupplier Diversity Partner SPDM128100UG
Antibody;100UG;Human IgG1/kappa;This product is a specific antibody against SARS-CoV-2 Spike protein RBD domain. No cross-reactivity is deteced with Spike protein RBD domain of other coronaviruses, including SARS-CoV, MERS-CoV, HCoV-229E, HCoV-NL63, HCoV-OC43 and HCoV-HKU1.;Anti-SARS-CoV-2 RBD Potent Neutralizing Antibody, Chimeric mAb, Human IgG1 (AM128) (SPD-M128) is a chimeric monoclonal antibody combining the constant domains of the human IgG1 molecule with mouse variable regions. Pseudovirus assay shows that this antibody has potent neutralizing activity against pseudovirus bearing SARS-CoV-2 Spike protein.;This protein . ;Immobilized SARS-CoV-2 S protein RBD, His Tag (Cat. No. SPD-C52H1) at 1 μg/mL (100 μL/well) can bind Anti-SARS-CoV-2 RBD Potent Neutralizing Antibody, Chimeric mAb, Human IgG1 (AM128) (Cat. No. SPD-M128) with a linear range of 0.2-1.5ng/mL (QC tested).;Serial dilutions of Anti-SARS-CoV-2 RBD Potent Neutralizing Antibody, Chimeric mAb, Human
Catalog No. 50-205-5036