Learn More
ACROBiosystems Human Syndecan-1 Protein, His Tag (MALS verified)
Supplier:  ACROBiosystems SY1H5225100UG
Encompass_Preferred
Recombinant Protein;100UG;Human Syndecan-1, His Tag (SY1-H5225) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Gly 254 (Accession # P18827-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.8 kDa. The protein migrates as 40-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Syndecan-1, His Tag (Cat. No. SY1-H5225) at 1 μg/mL (100 μL/well) can bind Anti-Syndecan-1 antibody, Mouse IgG1 (Cat. No. Anti-Syndecan-1 antibody) with a linear range of 0.1-3 ng/mL (QC tested).;Immobilized Anti-Syndecan-1 antibody, Mouse IgG1 (Cat. No. Anti-Syndecan-1 antibody) at 1 μg/mL (100 μL/well) can bind Human Syndecan-1, His Tag (Cat. No. SY1-H5225) with a linear range of 2-125 ng/mL (Routinely tested).
Catalog No. 50-205-5042