Learn More
ACROBiosystemsSupplier Diversity Partner Human PLGF / PGF (19-149) Protein, His Tag (MALS verified)
Manufacturer:  ACROBiosystemsSupplier Diversity Partner PGFH52H520UG
Encompass_Preferred
Recombinant Protein;20UG;Human PLGF (19-149) Protein, His Tag (PGF-H52H5) is expressed from human 293 cells (HEK293). It contains AA Leu 19 - Arg 149 (Accession # P49763-2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 16.6 kDa.
Catalog No. 50-205-5085