Learn More
ACROBiosystemsSupplier Diversity Partner Human Her2 / ErbB2 (S310F) Protein, His Tag (MALS verified)  DFS Item
Manufacturer:  ACROBiosystemsSupplier Diversity Partner HE2H52H2100UG
Recombinant Protein;100UG;Human Her2 (S310F), His Tag (HE2-H52H2) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 652 (Accession # P04626-1 (S310F)).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 71.3 kDa. The protein migrates as 80-100 KDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-205-5087