Learn More
ACROBiosystems Biotinylated Human CD3 epsilon&CD3 delta Heterodimer Protein, Fc,His,Avitag™&Fc,Flag,Avitag™ (MALS verified)
Supplier:  ACROBiosystems CDDH82F625UG
Encompass_Preferred
Recombinant Protein;25UG;MABSol Biotinylated Human CD3E&CD3D Heterodimer Protein, Fc,His,Avitag&Fc,Flag,Avitag (CDD-H82F6) is expressed from human HEK293 cells. It contains AA Asp 23 - Asp 126 (CD3E) & Phe 22 - Ala 105 (CD3D) (Accession # P07766-1 (CD3E) & P04234-1 (CD3D)). It is the biotinylated form of Human CD3 epsilon & CD3 delta Protein.;Biotinylated Human CD3E&CD3D Heterodimer Protein, Fc,His,Avitag&Fc,Flag,Avitag is produced by co-expression of CD3E and CD3D, has a calculated MW of 45.1kDa (CD3E) & 42.6 kDa (CD3D). Subunit CD3E is fused with a human IgG1 Fc fragment at the C-terminus, followed by an Avi tag (Avitag™), the polyhistidine tag is inserted in-between the Fc and Avi tags at the C-terminus and subunit CD3D contains a human IgG1 Fc fragment at the C-terminus, followed by an Avi tag (Avitag™), the flag tag is inserted in-between the Fc and Avi tags at the C-terminus.
Catalog No. 50-205-5093