Learn More
ACROBiosystems Mouse Anti-Human IgG2 secondary antibody, Mouse IgG1, HRP
Manufacturer:  ACROBiosystems IG2S7225UG
Encompass_Preferred
Antibody;25UG;IgG1/kappa;Does not cross-react with Human IgG1, IgG3 or IgG4.;Mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP2/0 myeloma and B-lymphocytes obtained from a mouse immunized with IgG2 F(ab')2.;This protein .
Catalog No. 50-205-5101