Learn More
ACROBiosystemsSupplier Diversity Partner Biotinylated Mouse FAP Protein, His,Avitag™
Manufacturer:  ACROBiosystemsSupplier Diversity Partner FAPM82Q525UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Mouse FAP, His,Avitag™ (FAP-M82Q5) is expressed from human 293 cells (HEK293). It contains AA Leu 26 - Asp 761 (Accession # P97321).;This protein carries a polyhistidine tag at the N-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 88.8 kDa. The protein migrates as 75-85 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Mouse FAP, His,Avitag (Cat. No. FAP-M82Q5) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate. can bind Monoclonal Anti-FAP (FAP5) Antibody, Mouse IgG2a with a linear range of 0.3-2 ng/mL (QC tested).;Immobilized Monoclonal Anti-FAP (FAP5) Antibody, Mouse IgG2a at 1 μg/mL (100 μL/well) can bind Biotinylated Mouse FAP, His,Avitag (Cat. No. FAP-M82Q5) with a linear range of 5-78 ng/mL (Routinely tested).
Catalog No. 50-205-5121