Learn More
ACROBiosystems Human NOTCH2 Protein, Fc Tag (MALS verified)
Supplier:  ACROBiosystems NO2H5255100UG
Encompass_Preferred
Recombinant Protein;100UG;Human NOTCH2, Fc Tag (NO2-H5255) is expressed from human 293 cells (HEK293). It contains AA Leu 26 - Gln 530 (Accession # Q04721-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 80.5 kDa. The protein migrates as 90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human DLL4, His Tag (Cat. No. DL4-H5227) at 5 μg/mL (100 μL/well) can bind Human NOTCH2, Fc Tag (Cat. No. NO2-H5255) with a linear range of 5-78 ng/mL (QC tested).;Immobilized Human NOTCH2, Fc Tag (Cat. No. NO2-H5255) at 1 μg/mL (100 μL/well) can bind Bispecific Antibody (NOTCH2 × NOTCH3) with a linear range of 1-20 ng/mL (Routinely tested).
Catalog No. 50-205-5139