Learn More
ACROBiosystems Biotinylated SARS-CoV-2 (COVID-19) S protein RBD, His,Avitag™ (MALS verified)
Supplier:  ACROBiosystems SPDC82E8200UG
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated SARS-CoV-2 S protein RBD, His,Avitag (SPD-C82E8) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Phe 541 (Accession # QHD43416.1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 28.2 kDa. The protein migrates as 33-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human ACE2, Fc Tag (Cat. No. AC2-H5257) at 2 μg/mL (100 μL/well) can bind Biotinylated SARS-CoV-2 S protein RBD, His,Avitag (Cat. No. SPD-C82E8) with a linear range of 10-313 ng/mL (QC tested).
Catalog No. 50-205-5145