Learn More
ACROBiosystems FITC-Labeled Human IL-3 R alpha / CD123 Protein, His Tag
Supplier:  ACROBiosystems ILAHF2H925UG
Encompass_Preferred
Recombinant Protein;25UG;FITC-Labeled Human IL-3 R alpha, His Tag (ILA-HF2H9) is expressed from human 293 cells (HEK293). It contains AA Thr 19 - Arg 305  (Accession # P26951-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 35.0 kDa.
Catalog No. 50-205-5172