Learn More
ACROBiosystems Biotinylated Human PVRIG Protein, Fc,Avitag™ (HPLC verified)
Supplier:  ACROBiosystems PVGH82F925UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human PVRIG, Fc,Avitag™ (HPLC verified) (PVG-H82F9) is expressed from human 293 cells (HEK293). It contains AA Thr 41 - Asp 171 (Accession # Q6DKI7-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 41.9 kDa. The protein migrates as 50-58 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Nectin-2, Mouse IgG2a Fc Tag (Cat. No. CD2-H5257) at 10 μg/mL (100 μL/well) can bind Biotinylated Human PVRIG, Fc,Avitag (HPLC verified) (Cat. No. PVG-H82F9) with a linear range of 5-40 ng/mL (QC tested).
Catalog No. 50-205-5192