Learn More
ACROBiosystemsSupplier Diversity Partner Human TREM2 Protein, His Tag  DFS Item
Manufacturer:  ACROBiosystemsSupplier Diversity Partner TR2H52H5100UG
Recombinant Protein;100UG;Human TREM2, His, Tag (TR2-H52H5) is expressed from human 293 cells (HEK293). It contains AA His 19 - Ser 174 (Accession # Q9NZC2-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 19.3 kDa. The protein migrates as 26-40 kDa under non-reducing (NR) condition (SDS-PAGE) due to glycosylation.;Immobilized Human TREM2, His, Tag (Cat. No. TR2-H52H5) at 1 μg/mL (100 μL/well) can bind Anti-TREM2 Antibody, Human IgG1 with a linear range of 0.3-2 ng/mL (QC tested).
Catalog No. 50-205-5196