Learn More
ACROBiosystemsSupplier Diversity Partner SARS-CoV-2 (COVID-19) S1 protein NTD (N234Q), His Tag
Manufacturer:  ACROBiosystemsSupplier Diversity Partner S1DC52H7100UG
Encompass_Preferred
Recombinant Protein;100UG;SARS-CoV-2 S1 protein NTD (N234Q), His Tag (S1D-C52H7) is expressed from human 293 cells (HEK293). It contains AA Ser 13 - Leu 303(N234Q) (Accession # QHD43416.1(N234Q)).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 34.9 kDa. The protein migrates as 45-65 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.;Immobilized SARS-CoV-2 S1 protein NTD (N234Q), His Tag (Cat. No. S1D-C52H7) at 1 μg/mL (100 μL/well) can bind Anti-SARS-CoV-2 Spike NTD Antibody, Chimeric mAb (Cat. No. SPD-M121) with a linear range of 0.2-6 ng/mL (QC tested).
Catalog No. 50-205-5202