Learn More
ACROBiosystemsSupplier Diversity Partner Biotinylated Onchocerca volvulus OV16 protein, Avi,GST Tag  DFS Item
Manufacturer:  ACROBiosystemsSupplier Diversity Partner OV6V81G925UG
Recombinant Protein;25UG;Biotinylated Onchocerca volvulus OV16 protein, Avi,GST Tag (OV6-V81G9) is expressed from E.coli cells. It contains AA Lys 17 - Asp 197 (Accession # P31729-1).;This protein carries an Avi tag at the N-terminus, followed by a GST tag. The protein has a calculated MW of 48.6 kDa. The protein migrates as 46-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-205-5244