Learn More
ACROBiosystems Human HGF Protein, His Tag (MALS verified)
Supplier:  ACROBiosystems HGFH52H3100UG
Encompass_Preferred
Recombinant Protein;100UG;Human HGF, His Tag (HGF-H52H3) is expressed from human 293 cells (HEK293). It contains AA Gln 32 - Ser 728 (Accession # P14210-1).;This protein carries a polyhistidine tag at the C-terminus. The mature form of HGF is a disulfide-linked heterodimer composed of proteolytically cleaved α and β chain. The protein has a calculated MW of 81.6 kDa (α chain 53.7 kDa and β chain 27.9 kDa). The protein migrates as 80-90kDa (alpha & Beta chain), 58-63 (alpha chain), 33 kDa (Beta chain) under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human HGF, His Tag at 2 μg/mL (100 μL/well) can bind Human HGF R, Fc Tag (Cat. No. MET-H5256) with a linear range of 1-39 ng/mL (QC tested).;Human HGF R Protein, Fc Tag (Cat. No. MET-H5256) captured on CM5 chip via Anti-human IgG Fc antibodies surface can bind Human HGF Protein, His Tag (Cat. No. HGF-H52H3) with an affinity constant of 75.2 pM as determined in a SPR assay (Biacore 8K) (QC tested).
Catalog No. 50-205-5311