Learn More
Sino Biological Mouse ALK-1 / ACVRL1 Protein (His & Fc Tag) 200ug
Supplier:  Sino Biological 50208M03H200
Encompass_Preferred
A DNA sequence encoding the mouse ALK1 (NP_033742.2) precursor (Met 1-Pro 119) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-1578
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.