Learn More
ACROBiosystems E1A SV40LTA DNA 100tests
Supplier:  ACROBiosystems OPAR007100TESTS
Encompass_Preferred
Residual host cell DNA refers to fragments of DNA from the host organism that are left behind after a biological process such as the production of a biopharmaceutical product or the cultivation of a cell line. This residual DNA can potentially contaminate the final product and affect its safety and efficacy.In the context of biopharmaceutical production regulatory agencies such as the FDA and EMA have set limits on the amount of residual host cell DNA that is acceptable in a final product. These limits vary depending on the type of product and the route of administration and residue host cell DNA quantitative kits are designed to ensure that the final product is safe for human use.
Catalog No. 50-253-4348
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.