Learn More
ACROBiosystems Cynomolgus KLRG1 100ug
Supplier:  ACROBiosystems KL1C5269100UG
Encompass_Preferred
Cynomolgus KLRG1 Protein Fc Tag (KL1-C5269) is expressed from human 293 cells (HEK293). It contains AA Leu 60 - Pro 189 (Accession # A0A2K5WAP9).
Catalog No. 50-253-4540
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.