Learn More
ACROBiosystems Mouse Cetuximab 100ug
Supplier:  ACROBiosystems CEBY29C100UG
Encompass_Preferred
Anti-Cetuximab Antibody (12B1E9) (CEB-Y29c) is a monoclonal antibody recombinantly expressed from human 293 cells (HEK293). The mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP2/0 myeloma and B-lymphocytes obtained from a mouse immunized with Cetuximab ADA.
Catalog No. 50-253-5294
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.