Learn More
PHCbi TwinGuard Series Ultra-Low Temperature Upright Freezer
Supplier: PHCbi MDFDU502VXCPA
PHCbi brand ULT freezers are engineered from the inside out to create, maintain, and restore precise temperatures necessary for vaccine storage. Our purpose-built ultra-low temperature freezers are designed for temperature uniformity and stability not commonly found in household or commercial cabinets.The MDF-DU502VXC-PA ultra-low temperature space-saving upright freezer provides reliable temperatures as low as -80°C. PHCbi brand TwinGuard ultra-low temperature freezers provide a wider circle of specimen protection through redundant cooling systems based on patented auto-cascade refrigeration technology.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.