Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Spectrum™ Labs Spectra/Por™ 5 12-14 kD MWCO Standard RC Dry Dialysis Kits
With Standard Grade Regenerated Cellulose membrane tubing for applications with significant size difference between MW species being separated
$220.00 - $228.00
Specifications
MWCO | 12 to 14 kD |
---|
Includes:
- 1m or 5m of Spectra/Por 5 dialysis tubing
- 1 Standard Spectra/Por Closure (for buoyance)
- 1 weighted Spectra/Por Closure (for orientation)
- 2 adhesive tubing openers
Catalog Number | Mfr. No. | Length (Metric) | Diameter (Metric) | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (Metric) | Diameter (Metric) | Quantity | Price | Quantity & Availability | |||
08-700-168
|
Spectrum™
132757T |
62 mL/cm vol./length | 89 mm | 5 m Tubing/Kit |
Each for $220.00
|
|
08-700-166
|
Spectrum™
132754T |
18 mL/cm vol./length | 48 mm | 5 m Tubing/Kit |
Each for $228.00
|
|
08-700-169
|
Spectrum™
132757T1 |
62 mL/cm vol./length | 89 mm | 1 m Tubing/Kit | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | View substitute products » |
08-700-167
|
Spectrum™
132754T1 |
18 mL/cm vol./length | 48 mm | 1 m Tubing/Kit | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | View substitute products » |
Description
- For reinforced for weight applications, or evaluating dialysis tubing for dialysis application
- Packaged dry without glycerin (no soaking required)
Specifications
12 to 14 kD |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title