Learn More
Cellpath System II Hex™ Tissue Processing Cassettes (Hoppered)
SDP
Pre-loaded into hoppers for use with automated and carousel type printers. CellPath System II Hex Tissue Processing Cassettes (Hoppered) feature unique honeycomb design that increases the open surface area compared to traditional round perforations, allowing increased reagent flow for improved processing and a more rigid block when sectioning.
Supplier:  Cellpath EAD010204H
 View more versions of this product
Catalog No. 22-222-074
Item Discontinued This product is either new or inactive, and cannot be ordered online.
Description
Description
  • Pre-loaded into hoppers for use with automated and carousel type cassette printers
  • Manufactured from high–grade virgin acetyl resin
  • Hexagonal perforations arranged in honeycombed structure to better utilize the available surface area
  • Unique arrangement increases open area thereby increasing reagent flow through and improving specimen staining and analysis time
  • Extra thick side walls increases rigidity and stability for high quality sectioning
  • Designed to suit Thermo Printmate and Micro-writer cassette–writing machines
  • Available in 15 stock colors which color code different types of workload or applications
Specifications
Specifications
1500/Pk.
White
Acetyl Resin
Tissue Processing Cassette in Hopper tube