Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Cellpath Promesh Safety Gloves
For better protection from the knife, scalpel and saw cuts. CellPath Promesh Safety Gloves consist of 10000 interlocking stainless steel that offers greater safety.
Manufacturer:  Cellpath XDA010400R
 View more versions of this product
Catalog No. 22-222-048
Description
Description
  • Features 10000 interlocking stainless steel links
  • Flexible and ambidextrous
Specifications
Specifications
Right
Medium
Red