Learn More
BioTek™ 50TS Microplate Washer Promo
Promo price is valid only with the purchase of one of the 800TS Microplate Readers.
Products 2
Catalog Number Mfr. No. Style Price Quantity & Availability  
Catalog Number Mfr. No. Style Price Quantity & Availability  
BT50TS8PM View Documents BioTek™
50TS8PM
96-Well This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
BT50TS8VPM View Documents BioTek™
50TS8VPM
96-Well, with Buffer Switching This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
For biomagnetic separation and vacuum filtration.
  • For 96-well washing
  • Automated
  • 12-well manifold
  • 4.3 in. color touchscreen LCD
  • 1 to 10 wash cycles
Warranty and Services
12 months