Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

58K Golgi Protein Mouse anti-Human, Mouse, DyLight 650, Clone: 575, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243573C

Catalog No. NBP243573C

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

58K Golgi Protein
575
Western Blot
Purified
FTCD
Recombinant protein encompassing a sequence within the C-terminus region of human FTCD. The exact sequence is proprietary.
Protein G purified
Cellular Markers, Golgi Apparatus Markers, Lipid and Metabolism, Phospho Specific, Signal Transduction
Primary
10841
Western Blot
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
formimidoyltransferase cyclodeaminase, formimidoyltransferase-cyclodeaminase, formiminotransferase cyclodeaminase, Formiminotransferase-cyclodeaminase, human formiminotransferase cyclodeaminase, EC 4.3.1.410formiminotransferase-cyclodeaminase, LCHC1
Mouse
IgG2b
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit