Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
58KGolgiProtein Rabbit anti-Human, Clone: JF099-5, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP267099
Description
58K Golgi Protein Monoclonal specifically detects 58K Golgi Protein in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
58KGolgiProtein | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin | |
formimidoyltransferase cyclodeaminase, formimidoyltransferase-cyclodeaminase, formiminotransferase cyclodeaminase, Formiminotransferase-cyclodeaminase, human formiminotransferase cyclodeaminase, EC 4.3.1.410formiminotransferase-cyclodeaminase, LCHC1 | |
Rabbit | |
Protein A purified | |
Cellular Markers, Golgi Apparatus Markers, Lipid and Metabolism, Phospho Specific, Signal Transduction | |
10841 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
JF099-5 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
FTCD | |
Synthetic peptide within Human 58K Golgi Protein aa 51-100/ 541. (SwissProt: O95954 Human) | |
100 μL | |
Primary | |
Human | |
Purified |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction