Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5H-Chromeno[2,3-b]pyridin-5-one, 97%, Maybridge™

Click to view available options
Quantity:
1g
5g
6537-46-8
C12H7NO2
MFCD00052253
6537-46-8
C12H7NO2
MFCD00052253
Quantity 1g
Packaging Amber glass bottle
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 197.19
Percent Purity 97%
Chemical Name or Material 5H-Chromeno[2, 3-b]pyridin-5-one