Learn More
Abnova™ 5q31.2 (Orange) FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Supplier:  Abnova™ FG0106
Catalog No. 89-932-022
Add to Cart
Description
Description
Specifications
Specifications
Store at -20°C in the dark.
FISH
200 μL
Product Suggestions
Product Suggestions