Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5Z-7-Oxozeaenol

Click to view available options
Quantity:
1mg
5mg
66018-38-0
C19H22O7
362.378
MFCD12912700
NEQZWEXWOFPKOT-UHFFFAOYSA-N
8,9,16-trihydroxy-14-methoxy-3-methyl-4,8,9,10-tetrahydro-3h-2-benzoxacyclotetradecine-1,7-dione
53442201
5,6,15-trihydroxy-17-methoxy-11-methyl-12-oxabicyclo[12.4.0]octadeca-1(14),2,8,15,17-pentaene-7,13-dione
CC1CC=CC(=O)C(C(CC=CC2=CC(=CC(=C2C(=O)O1)O)OC)O)O
66018-38-0
C19H22O7
362.378
MFCD12912700
NEQZWEXWOFPKOT-UHFFFAOYSA-N
8,9,16-trihydroxy-14-methoxy-3-methyl-4,8,9,10-tetrahydro-3h-2-benzoxacyclotetradecine-1,7-dione
53442201
5,6,15-trihydroxy-17-methoxy-11-methyl-12-oxabicyclo[12.4.0]octadeca-1(14),2,8,15,17-pentaene-7,13-dione
CC1CC=CC(=O)C(C(CC=CC2=CC(=CC(=C2C(=O)O1)O)OC)O)O
Color White
Quantity 1mg
UN Number UN2811
Solubility Information Soluble in DMSO
Formula Weight 362.37
Physical Form Solid
Chemical Name or Material 5Z-7-Oxozeaenol

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

storageNote1 : -30° to -10°C