Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6,2',4'-Trimethoxyflavone, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
720675-74-1
C18H16O5
312.321
WUWFDVDASNSUKP-UHFFFAOYSA-N
6,2',4'-trimethoxyflavone, 2-2,4-dimethoxyphenyl-6-methoxychromen-4-one, 2-2,4-dimethoxyphenyl-6-methoxy-4h-chromen-4-one, flavonid, 6 4,2, 2',4',6-trimethoxyflavone, 6,2/',4/'-trimethoxyflavone, 6, 2', 4'-trimethoxyflavone hplc, 4h-1-benzopyran-4-one, 2-2,4-dimethoxyphenyl-6-methoxy, 2-2,4-dimethoxyphenyl-2,3-dihydro-6-methoxy-4h-1-benzopyran-4-one, 6, 2 inverted exclamation marka, 4 inverted exclamation marka-trimethoxyflavone
688802
2-(2,4-dimethoxyphenyl)-6-methoxychromen-4-one
COC1=CC2=C(C=C1)OC(=CC2=O)C3=C(C=C(C=C3)OC)OC
This item is not returnable. View return policy
720675-74-1
C18H16O5
312.321
WUWFDVDASNSUKP-UHFFFAOYSA-N
6,2',4'-trimethoxyflavone, 2-2,4-dimethoxyphenyl-6-methoxychromen-4-one, 2-2,4-dimethoxyphenyl-6-methoxy-4h-chromen-4-one, flavonid, 6 4,2, 2',4',6-trimethoxyflavone, 6,2/',4/'-trimethoxyflavone, 6, 2', 4'-trimethoxyflavone hplc, 4h-1-benzopyran-4-one, 2-2,4-dimethoxyphenyl-6-methoxy, 2-2,4-dimethoxyphenyl-2,3-dihydro-6-methoxy-4h-1-benzopyran-4-one, 6, 2 inverted exclamation marka, 4 inverted exclamation marka-trimethoxyflavone
688802
2-(2,4-dimethoxyphenyl)-6-methoxychromen-4-one
COC1=CC2=C(C=C1)OC(=CC2=O)C3=C(C=C(C=C3)OC)OC

Aryl hydrocarbon receptor antagonist

Aryl hydrocarbon receptor antagonist (EC50 = 0.9 μM). Exhibits no short term agonist activity and no species or promoter dependence.
Quantity 10mg
Formula Weight 312.32
Percent Purity >97%
Chemical Name or Material 6, 2', 4'-Trimethoxyflavone

Recommended Storage : Store at Room Temperature