Alfa Aesar™ 6-(4-Ethoxycarbonyl-1-piperazinyl)pyridine-3-boronic acid pinacol ester, 95%

Manufacturer: Alfa Aesar™ H2712203

Quantity 1g

 View more versions of this product

Catalog No. AAH2712203

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1073354-26-3
    Molecular Formula C18H28BN3O4
    Formula Weight 361.25
    MDL Number MFCD06798269
    Synonym 4-[5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-pyridyl]piperazine-1-carboxylic acid ethyl ester
    Chemical Name or Material 6-(4-Ethoxycarbonyl-1-piperazinyl)pyridine-3-boronic acid pinacol ester

    Specifications

    Formula Weight 361.25
    Molecular Formula C18H28BN3O4
    MDL Number MFCD06798269
    Synonym 4-[5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-pyridyl]piperazine-1-carboxylic acid ethyl ester
    Melting Point 135° to 139°C
    Quantity 1g
    CAS 1073354-26-3
    Chemical Name or Material 6-(4-Ethoxycarbonyl-1-piperazinyl)pyridine-3-boronic acid pinacol ester
    Assay 95%

    Description

    Applications
    6-(4-Ethoxycarbonyl-1-piperazinyl)pyridine-3-boronic acid pinacol ester, is used as a main benefits of the use of boronic esters in Suzuki-Miyaura cross-couplings- Enabling the coupling of organoboronic acids that are sensitive to hydrolytic deboronation. It includes other additional application such as a number of chiral boronic esters, such as the ones illustrated hereafter, have been used as inducers in stereoselective reactions.

    Solubility
    Insoluble in water. Soluble in organic solvents.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit