Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-(4-Fluorophenyl)-2-pyridinecarboxaldehyde, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
157402-44-3
C12H9NO
183.21
PVZMGWZOXQPFLS-UHFFFAOYSA-N
6-phenylpicolinaldehyde,6-phenylpyridine-2-carboxaldehyde,2-pyridinecarboxaldehyde, 6-phenyl,6-phenyl-pyridine-2-carbaldehyde,2-formyl-6-phenylpyridine,6-phenyl-2-pyridine aldehyde,6-phenyl-2-pyridinecarboxaldehyde
13522416
6-phenylpyridine-2-carbaldehyde
C1=CC=C(C=C1)C2=NC(=CC=C2)C=O
157402-44-3
C12H9NO
183.21
PVZMGWZOXQPFLS-UHFFFAOYSA-N
6-phenylpicolinaldehyde,6-phenylpyridine-2-carboxaldehyde,2-pyridinecarboxaldehyde, 6-phenyl,6-phenyl-pyridine-2-carbaldehyde,2-formyl-6-phenylpyridine,6-phenyl-2-pyridine aldehyde,6-phenyl-2-pyridinecarboxaldehyde
13522416
6-phenylpyridine-2-carbaldehyde
C1=CC=C(C=C1)C2=NC(=CC=C2)C=O
Quantity 5g
Assay Percent Range 97%
Formula Weight 201.2
Chemical Name or Material 6-(4-Fluorophenyl)-2-pyridinecarboxaldehyde