Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6,6'-Dibromo-1,1'-bi-2-naphthol, 95%, Thermo Scientific™

Catalog No. AC350490250
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13185-00-7
Molecular Formula C20H12Br2O2
Molecular Weight (g/mol) 444.13
MDL Number MFCD00798290
InChI Key OORIFUHRGQKYEV-UHFFFAOYSA-N
Synonym r---6,6'-dibromo-1,1'-bi-2-naphthol, s---6,6'-dibromo-1,1'-bi-2-naphthol, s-+-6,6'-dibromo-1,1'-bi-2-naphthol, 6,6'-dibromo-1,1'-bi-2-naphthol, s-+-6,6'-dibromo-1,1'-binaphthalene-2,2'-diol, r---6,6'-dibromo-1,1'-binaphthalene-2,2'-diol, 6,6'-dibromo 1,1'-binaphthalene-2,2'-diol, +/--6,6'-dibromo-1,1'-bi-2-naphthol, s-6,6'-dibromo-1,1'-bi-2-naphthol, racemic-6,6'-dibromo-1,1'-bi-2-naphthol
PubChem CID 222842
IUPAC Name 6-bromo-1-(6-bromo-2-hydroxynaphthalen-1-yl)naphthalen-2-ol
SMILES C1=CC2=C(C=CC(=C2C3=C(C=CC4=C3C=CC(=C4)Br)O)O)C=C1Br

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 201°C to 207°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: insoluble
Formula Weight 444.13
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 6, 6'-Dibromo-1, 1'-bi-2-naphthol, 95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.