Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6,6-Dimethyl-3-azabicyclo[3.1.0]hexane-2,4-dione, 98%, Alfa Aesar™

Click to view available options
Quantity:
1g
5g
194421-56-2
C7H9NO2
139.154
MFCD12923551
DRMNZTFLOOSXIN-UHFFFAOYSA-N
6,6-dimethyl-3-azabicyclo 3.1.0 hexane-2,4-dione,3,3-dimethylcyclopropane-1,2-dicarboximide,3-azabicyclo 3.1.0 hexane-2,4-dione, 6,6-dimethyl,3-azabicyclo 3.1.0 hexane-2,4-dione,6,6-dimethyl
10749252
6,6-dimethyl-3-azabicyclo[3.1.0]hexane-2,4-dione
CC1(C2C1C(=O)NC2=O)C
194421-56-2
C7H9NO2
139.154
MFCD12923551
DRMNZTFLOOSXIN-UHFFFAOYSA-N
6,6-dimethyl-3-azabicyclo 3.1.0 hexane-2,4-dione,3,3-dimethylcyclopropane-1,2-dicarboximide,3-azabicyclo 3.1.0 hexane-2,4-dione, 6,6-dimethyl,3-azabicyclo 3.1.0 hexane-2,4-dione,6,6-dimethyl
10749252
6,6-dimethyl-3-azabicyclo[3.1.0]hexane-2,4-dione
CC1(C2C1C(=O)NC2=O)C
Quantity 1g
Beilstein 117393
Formula Weight 139.15
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 6,6-Dimethyl-3-azabicyclo[3.1.0]hexane-2,4-dione